Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1S Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA530102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530102 Supplier Invitrogen™ Supplier No. MA530102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Complement C1s.

This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1S
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% trehalose, 0.02% Tween 80 and no preservative; pH 7.0
Gene C1S
Gene Accession No. P09871
Gene Alias AA959438; AI255193; AI327365; basic proline-rich peptide IB-1; C1 esterase; C1s; C1s1; C1sa; complement C1s; complement C1s subcomponent; Complement C1s subcomponent heavy chain; Complement C1s subcomponent light chain; Complement C1s-A subcomponent; Complement C1s-A subcomponent heavy chain; Complement C1s-A subcomponent light chain; Complement component 1 subcomponent s; complement component 1 subcomponent s-A; complement component 1, s subcomponent; complement component 1, s subcomponent 1; complement component C1SA; r-gsp; serine protease
Gene Symbols C1S
Host Species Mouse
Immunogen Recombinant Human Complement C1s Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 716
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.