Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1S Monoclonal Antibody (9)
GREENER_CHOICE

Catalog No. ABS0020902
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0020902 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0020902 Supplier Invitrogen™ Supplier No. ABS0020902
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 002-09-02 is specific for C1s proenzyme and activated free C1s. ABS 002-09-02 has been successfully used in ELISA procedures. ABS 002-09-02 reacts in ELISA when tested with activated C1s enzyme directly coated onto the microtiter well or in sandwich ELISA in combination with a polyclonal antibody in coat. Epitope of ABS 002-09-02 differs from that of ABS 002-49-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1S
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene C1S
Gene Accession No. P09871
Gene Alias AA959438; AI255193; AI327365; basic proline-rich peptide IB-1; C1 esterase; C1s; C1s1; C1sa; complement C1s; complement C1s subcomponent; Complement C1s subcomponent heavy chain; Complement C1s subcomponent light chain; Complement C1s-A subcomponent; Complement C1s-A subcomponent heavy chain; Complement C1s-A subcomponent light chain; Complement component 1 subcomponent s; complement component 1 subcomponent s-A; complement component 1, s subcomponent; complement component 1, s subcomponent 1; complement component C1SA; r-gsp; serine protease
Gene Symbols C1S
Host Species Mouse
Immunogen C1s proenzyme purified from human plasma.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 716
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.