Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C1S Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582638 Supplier Invitrogen™ Supplier No. PA582638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAHVVEGNRE PTMYVGSTSV QTSRLAKSKM LTPEHVFIHP GWKLLEVPEG RTNFDNDIAL VRLKDPVKMG PTVSPICLPG TSSDYNLMDG DLGLISGWGR TEKRDRAVRL KAARLPVAPL RKCKEVKVEK PTADAEAYVF TPNMICAGGE Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 61%.

This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1S
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1S
Gene Accession No. P09871
Gene Alias AA959438; AI255193; AI327365; basic proline-rich peptide IB-1; C1 esterase; C1s; C1s1; C1sa; complement C1s; complement C1s subcomponent; Complement C1s subcomponent heavy chain; Complement C1s subcomponent light chain; Complement C1s-A subcomponent; Complement C1s-A subcomponent heavy chain; Complement C1s-A subcomponent light chain; Complement component 1 subcomponent s; complement component 1 subcomponent s-A; complement component 1, s subcomponent; complement component 1, s subcomponent 1; complement component C1SA; r-gsp; serine protease
Gene Symbols C1S
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1S. Recombinant protein control fragment (Product #RP-95390).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 716
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.