Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C2 Mouse anti-Human, Clone: OTI1A7, lyophilized, TrueMAB™
SDP

Catalog No. 501668137
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8137 Supplier OriGene Supplier No. CF807163
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Component C2 is a serum glycoprotein that functions aspart of the classical pathway of the complement system. ActivatedC1 cleaves C2 into C2a and C2b. The serine proteinase C2a thencombines with complement factor 4b to create the C3 or C5convertase. Deficiency of C2 has been reported to associated withcertain autoimmune diseases and SNPs in this gene have beenassociated with altered susceptibility to age-related maculardegeneration. This gene localizes within the class III region ofthe MHC on the short arm of chromosome 6. Alternative splicingresults in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described in publicationsbut their full-length sequence has not been determined.

Specifications

Antigen C2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A7
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene C2
Gene Accession No. P06681
Gene Alias ARMD14, CO2
Gene Symbols C2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 469-752 of human C2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 717
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.