Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544101 Supplier Invitrogen™ Supplier No. PA544101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RLRTVLKNLS LLKLLKSSNR RIQELHKLAK RCWHSLLSVP KILRISSGEN Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 83%; Human: 100%; Mouse: 93%; Pig: 86%; Rabbit: 79%; Rat: 86%.

May play a significant role in p53/TP53-mediating signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TP53TG5
Gene Accession No. Q9Y2B4
Gene Alias C20orf10; CLG01; P3H3; RGD1561934; TP53 target 5; TP53-inducible gene 5 protein; TP53-target gene 5 protein; TP53TG5
Gene Symbols TP53TG5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C20orf10 (aa 38-87).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27296
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.