Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf103 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9266 Supplier Bioss Supplier No. BS9691R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysosome-associated membrane protein 5 (LAMP5) is a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.

Specifications

Antigen C20orf103
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LAMP5
Gene Accession No. Q9UJQ1
Gene Alias 3110035N03Rik; 6330527O06Rik; BADLAMP; BAD-LAMP; BOS_12751; brain and dendritic cell associated LAMP; Brain and dendritic cell-associated LAMP; Brain-associated LAMP-like protein; C13H20orf103; C20orf103; C24H20orf103; chromosome 20 open reading frame 103; LAMP family protein C20orf103; LAMP family protein C20orf103 homolog; Lamp5; LAMP-5; LOC100340997; lysosomal associated membrane protein family member 5; lysosomal-associated membrane protein family, member 5; Lysosome-associated membrane glycoprotein 5; Lysosome-associated membrane protein 5; Protein C20orf103; RGD1306991; UNC-46; UPF0710 protein C20orf103 homolog; zgc:165600
Gene Symbols LAMP5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf103.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24141
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.