Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C20orf107 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559888
Description
Immunogen sequence: FPFRTPLKKN QNASLYKDCV FNTLN Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.
Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20.
Specifications
C20orf107 | |
Polyclonal | |
Unconjugated | |
FAM209B | |
C20orf107; dJ1153D9.4; FAM209B; family with sequence similarity 209 member B; family with sequence similarity 209, member B; protein FAM209B; uncharacterized protein C20orf107 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JX69 | |
FAM209B | |
Recombinant protein corresponding to Human C20orf107. Recombinant protein control fragment (Product #RP-99288). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction