Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf111 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989267
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9267 Supplier Bioss Supplier No. BS9692R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C20orf111
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene OSER1
Gene Accession No. Q9NX31
Gene Alias BM-038; C20orf111; dJ1183I21.1; HSPC168; HSPC207; OSER1; Osr1; oxidative stress responsive serine rich 1; oxidative stress responsive serine-rich 1; oxidative stress-responsive 1; Oxidative stress-responsive protein 1; Oxidative stress-responsive serine-rich protein 1; Perit1; Peroxide-inducible transcript 1 protein
Gene Symbols OSER1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf111.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51526
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.