Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf114 Mouse anti-Human, Clone:2A5, Abnova™

Catalog No. 89334564 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-564 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-564 Supplier Abnova Corporation Supplier No. MAB17709
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human C20orf114.

The protein encoded by this gene may be involved in the innate immune response to bacterial exposure in the mouth, nasal cavities, and lungs. The encoded protein is secreted and is a member of the BPI/LBP/PLUNC protein superfamily. This gene is found with other members of the superfamily in a cluster on chromosome 20. [provided by RefSeq]

Specifications

Antigen C20orf114
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2A5
Conjugate Unconjugated
Description chromosome 20 open reading frame 114
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry (1:200-1:1000)
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene C20orf114
Gene Alias LPLUNC1, MGC14597
Host Species Mouse
Immunogen Recombinant protein corresponding to human C20orf114 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 92747
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.