Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563734 Supplier Invitrogen™ Supplier No. PA563734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKMSDHKPLL TAISPGRGYW RRQLDHISAH LRCYAQNNPE FRALIAEPRM GKLISATVHE DGCEVSIRLN YSQVSNKNLY LNKAVNFSDH LLSSAA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

This gene contains two ankyrin repeat-encoding regions. Ankyrin repeats are tandemly repeated modules of about 33 amino acids described as L-shaped structures consisting of a beta-hairpin and two alpha-helices. Ankyrin repeats occur in a large number of functionally diverse proteins, mainly from eukaryotes, and are known to function as protein-protein interaction domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DZANK1
Gene Accession No. Q9NVP4
Gene Alias ANKRD64; ankyrin repeat domain 64; ankyrin repeat-containing protein C20orf12; bA189K21.1; bA189K21.8; C20orf12; C20orf84; dJ568F9.2; double zinc ribbon and ankyrin repeat domains 1; double zinc ribbon and ankyrin repeat-containing protein 1; DZANK1
Gene Symbols DZANK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C20orf12. Recombinant protein control fragment (Product #RP-99314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.