Learn More
Invitrogen™ C20orf132 Polyclonal Antibody

Description
Immunogen sequence: EPLSHSPRHL TQQDPLSEAI VEKLIQSIQK VFNGELKGEL EKLKFLGDLS SLSQALPHDE TAKSFIHSHI ADIVHTLNVL VQEERPHSLS SSMRQEVFVT IAD Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.
Specifications
Specifications
| Antigen | C20orf132 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MROH8 |
| Gene Accession No. | Q9H579 |
| Gene Alias | 4922505G16Rik; C20orf131; C20orf132; dJ621N11.3; dJ621N11.4; maestro heat like repeat family member 8; maestro heat-like repeat family member 8; maestro heat-like repeat-containing protein family member 8; MROH8; Protein MROH8 |
| Gene Symbols | MROH8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.