Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C20orf14 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15094R

Description
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.Specifications
C20orf14 | |
Polyclonal | |
Unconjugated | |
PRPF6 | |
1190003A07Rik; 2610031L17Rik; androgen receptor N-terminal domain transactivating protein-1; Androgen receptor N-terminal domain-transactivating protein 1; ANT1; ANT-1; C20orf14; hPrp6; p102 U5 small nuclear ribonucleoprotein particle-binding protein; pre-mRNA processing factor 6; Pre-mRNA-processing factor 6; Prp6; PRP6 homolog; PRP6 pre-mRNA processing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog (yeast); Prpf6; putative mitochondrial outer membrane protein import receptor; RP60; SNRNP102; TOM; U5 snRNP-associated 102 kDa protein; U5-102 kDa protein; U5-102K | |
Rabbit | |
Protein A | |
RUO | |
24148 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O94906 | |
PRPF6 | |
KLH conjugated synthetic peptide derived from human C20orf14. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction