Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C20ORF141 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | C20ORF141 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
C20ORF141 Polyclonal specifically detects C20ORF141 in Human samples. It is validated for Western Blot.Specifications
| C20ORF141 | |
| Polyclonal | |
| Rabbit | |
| Q9NUB4 | |
| 128653 | |
| Synthetic peptides corresponding to C20ORF141 The peptide sequence was selected from the middle region of C20ORF141. Peptide sequence HTLPQRKLLTRGQSQGAGEGPGQQEALLLQMGTVSGQLSLQDALLLLLMG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| chromosome 20 open reading frame 141, dJ860F19.4, hypothetical protein LOC128653, MGC26144 | |
| C20ORF141 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title