Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf141 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543834 Supplier Invitrogen™ Supplier No. PA543834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RKLLTRGQSQ GAGEGPGQQE ALLLQMGTVS GQLSLQDALL LLLMGLGPLL Sequence homology: Human: 100%.

C20orf141 is a single-pass membrane protein. The exact function of C20orf141 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf141
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C20orf141
Gene Accession No. Q9NUB4
Gene Alias C20orf141; chromosome 20 open reading frame 141; dJ860F19.4; Uncharacterized protein C20orf141
Gene Symbols C20orf141
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C20orf141.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128653
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.