Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf160 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989270
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9270 Supplier Bioss Supplier No. BS9695R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCM2L gene ontology annotations related to this gene include cardiac atrium morphogenesis; cardiac muscle tissue growth; negative regulation of homotypic cell-cell adhesion; negative regulation of protein binding; positive regulation of fibroblast growth factor production; ventricular trabecula myocardium morphogenesis; wound healing.

Specifications

Antigen C20orf160
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ccm2l
Gene Accession No. Q9NUG4
Gene Alias C20orf160; C24H20orf160; CCM2 like scaffold protein; CCM2 like scaffolding protein; CCM2L; CCM2-like; ccml2; cerebral cavernous malformation 2-like; Cerebral cavernous malformations 2 protein-like; dJ310O13.5; LOW QUALITY PROTEIN: cerebral cavernous malformations 2 protein-like; RGD1305202; si:dkey-223d9.2
Gene Symbols Ccm2l
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf160.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140706
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.