Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566033 Supplier Invitrogen™ Supplier No. PA566033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPGRSKLIASA LSKPLPHQEG ASKSGPSRKK AFHHEEIHPS HYAFETSPRP IDVLGGTRFC SQRQTLEMRT EE Highest antigen sequence identity to the following orthologs - mouse 66%, rat 63%.

FAM217B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf177
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM217B
Gene Accession No. Q9NTX9
Gene Alias C20orf177; dJ551D2.5; FAM217B; family with sequence similarity 217 member B; family with sequence similarity 217, member B; protein FAM217B; uncharacterized protein C20orf177
Gene Symbols FAM217B
Host Species Rabbit
Immunogen Recombinant Human C20orf177. Recombinant protein control fragment (Product #RP-106722).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.