Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf177 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983455
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3455 Supplier Bioss Supplier No. BS15101R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM217B is a protein coding gene.

Specifications

Antigen C20orf177
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM217B
Gene Accession No. Q9NTX9
Gene Alias C20orf177; dJ551D2.5; FAM217B; family with sequence similarity 217 member B; family with sequence similarity 217, member B; Protein FAM217B; uncharacterized protein C20orf177
Gene Symbols FAM217B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf177.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63939
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.