Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf194 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540557 Supplier Invitrogen™ Supplier No. PA540557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SFAKHMVAQC VSPKGPLACS RTYFFGATHV PYLGGDSKLP KKTEQIRLLS Sequence homology: Cow: 86%; Human: 100%; Pig: 79%.

The function of the C20orf194 gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf194
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C20orf194
Gene Accession No. Q5TEA3
Gene Alias C20orf194; chromosome 20 open reading frame 194; RGD1565616; Uncharacterized protein C20orf194; uncharacterized protein C20orf194 homolog
Gene Symbols C20orf194
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C20orf194 (aa 322-371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25943
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.