Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf194 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562551 Supplier Invitrogen™ Supplier No. PA562551
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYCNPVNFRY LLPYVAHWRN LHFHCMTENE YEDEEAAEEF KITSFVDMVR DCSRIGIPYS SQGHLQIFDM FVVEKWPIVQ AFALEGIGGD GFFTMKYELQ DVSLN Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

The function of the C20orf194 gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf194
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C20orf194
Gene Accession No. Q5TEA3
Gene Alias C20orf194; chromosome 20 open reading frame 194; RGD1565616; Uncharacterized protein C20orf194; uncharacterized protein C20orf194 homolog
Gene Symbols C20orf194
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C20orf194. Recombinant protein control fragment (Product #RP-99299).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.