Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf195 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544121 Supplier Invitrogen™ Supplier No. PA544121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FDRKASAAHQ DWARLRWFVT IQPATSEQYE LRFRLLDPRT QQECAQCGVI Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%.

The exact function of C20orf195 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf195
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FNDC11
Gene Accession No. Q9BVV2
Gene Alias 1700026M20Rik; BC051628; C20orf195; cDNA sequence BC051628; fibronectin type III domain containing 11; Fibronectin type III domain-containing protein 11; Fndc11; RGD1564340; similar to hypothetical protein MGC5356; uncharacterized protein C20orf195; uncharacterized protein C20orf195 homolog
Gene Symbols FNDC11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C20orf195 (aa 212-261).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79025
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.