Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C20orf197 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571015
Description
C20orf197 Polyclonal Antibody for Western Blot
C20orf197 is a protein coding gene.
Specifications
C20orf197 | |
Polyclonal | |
Unconjugated | |
C20orf197 | |
C20orf197; chromosome 20 open reading frame 197; uncharacterized protein C20orf197 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
284756 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N268 | |
C20orf197 | |
synthetic peptide directed towards the N-terminal region of human C20orf197. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction