Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf202 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560119 Supplier Invitrogen™ Supplier No. PA560119
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYKSKIPRAQ NQVSVKVTPK NTEMKIAEEP SPSLGQTLEW LRKELSEMQI QDQSLLLTLR HL Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 27%.

C20orf202 is a 122 amino acid protein that is pending further characterization. The gene encoding C20orf202 maps to human chromosome 20. Comprising approximately 2% of the human genome, chromosome 20 contains nearly 63 million bases that encode over 600 genes, some of which are associated with Creutzfeldt-Jakob disease, amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome. Additionally, chromosome 20 contains a region with numerous genes which are thought to be important for seminal production and may be potential targets for male contraception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf202
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C20orf202
Gene Accession No. A1L168
Gene Alias C20orf202; chromosome 20 open reading frame 202; uncharacterized protein C20orf202
Gene Symbols C20orf202
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C20orf202. Recombinant protein control fragment (Product #RP-99483).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400831
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.