Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf203 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568728 Supplier Invitrogen™ Supplier No. PA568728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

The protein encoded by this gene is thought to be a human-specific protein. Currently available evidence suggests that orthologous regions in other organisms contain sequence differences that would not support production of a protein product. Genome-wide association studies have suggested the possibility that a SNP in the 3' UTR, rs17123507, could be associated with nicotine addiction. Expression of this gene may be elevated in some individuals with Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf203
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C20orf203
Gene Accession No. Q8NBC4
Gene Alias alugen; C20orf203; chromosome 20 open reading frame 203; uncharacterized protein C20orf203
Gene Symbols C20orf203
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human C20orf203.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284805
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.