Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB5IF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112105 Supplier Thermo Scientific Supplier No. PA5112105
Add to Cart
Add to Cart

C20orf24 Polyclonal Antibody

C20orf24 Polyclonal Antibody for ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5IF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene C20orf24
Gene Accession No. Q9BUV8
Gene Alias C20orf24; chromosome 20 open reading frame 24; PNAS-11; RAB5IF; Rab5-interacting protein; RIP5; uncharacterized protein C20orf24; Uncharacterized protein RAB5IF
Gene Symbols C20orf24
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human C20orf24/RAB5IF.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.