Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB5IF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543332 Supplier Invitrogen™ Supplier No. PA543332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSGGRRKEEP PQPQLANGAL KVSVWSKVLR SDAAWEDKDE FLDVIYWFRQ Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 90%.

RAB5IF, also known as RAB5-interacting factor, is a protein that interacts with the small GTPase RAB5 and plays a role in endocytic trafficking. It is involved in the regulation of early endosome fusion and recycling, and has been implicated in various cellular processes such as cell migration and cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5IF
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C20orf24
Gene Accession No. Q9BUV8
Gene Alias C20orf24; chromosome 20 open reading frame 24; PNAS-11; RAB5IF; Rab5-interacting protein; RIP5; uncharacterized protein C20orf24; Uncharacterized protein RAB5IF
Gene Symbols C20orf24
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C20orf24 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55969
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.