Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983460
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3460 Supplier Bioss Supplier No. BS15106R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function remain unknown.

Specifications

Antigen C20orf24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C20orf24
Gene Accession No. Q9BUV8
Gene Alias C20orf24; chromosome 20 open reading frame 24; PNAS-11; RAB5IF; Rab5-interacting protein; RIP5; uncharacterized protein C20orf24; Uncharacterized protein RAB5IF
Gene Symbols C20orf24
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55969
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.