Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf3 Monoclonal Antibody (OTI1A9), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504318

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-5037


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

C20orf3 may play a role in adipocyte differentiation.
Specifications

Specifications

C20orf3
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9HDC9
APMAP
Full length human recombit protein of human C20orf3 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1A9
Unconjugated
APMAP
BSCv, C20orf3
Mouse
Affinity Chromatography
RUO
57136
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.