Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf3 Monoclonal Antibody (OTI1D5), TrueMAB™, OriGene
SDP

Catalog No. 501675176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-5176 100 μL
50-254-0285 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-5176 Supplier OriGene Supplier No. TA504500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C20orf3 may play a role in adipocyte differentiation.

Specifications

Antigen C20orf3
Applications Western Blot
Classification Monoclonal
Clone OTI1D5
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene APMAP
Gene Accession No. Q9HDC9
Gene Alias BSCv, C20orf3
Gene Symbols APMAP
Host Species Mouse
Immunogen Full length human recombit protein of human C20orf3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57136
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.