Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf3 Monoclonal Antibody (OTI4F6)
GREENER_CHOICE

Catalog No. PIMA525848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525848 Supplier Invitrogen™ Supplier No. MA525848
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C20orf3 Monoclonal Antibody for IHC (P), Western Blot

APMAP (Adipocyte Plasma Membrane Associated Protein) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include arylesterase activity and strictosidine synthase activity. Exhibits strong arylesterase activity with beta-naphthyl acetate and phenyl acetate. May play a role in adipocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3
Gene APMAP
Gene Accession No. Q7TP48, Q9D7N9, Q9HDC9
Gene Alias Adipocyte plasma membrane-associated protein; APMAP; C20orf3
Gene Symbols APMAP
Host Species Mouse
Immunogen Full length human recombinant protein of C20orf3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366227, 57136, 608089, 71881
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.