Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C20orf30 Monoclonal Antibody (OTI1B4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504919

Description
Involved in trafficking and recycling of synaptic vesicles.Specifications
C20orf30 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96A57 | |
TMEM230 | |
Full length human recombit protein of human C20orf30 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI1B4 | |
Unconjugated | |
TMEM230 | |
C20orf30, HSPC274, dJ1116H23.2.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29058 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction