Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf30 Monoclonal Antibody (OTI3B2), TrueMAB™, OriGene
SDP

Catalog No. 502540019
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0019 30 μL
50-167-5523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0019 Supplier OriGene Supplier No. TA504942S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C20orf30 Monoclonal antibody specifically detects C20orf30 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen C20orf30
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LRRC25
Gene Accession No. Q8N386
Gene Alias 5730494N06Rik; AA407821; C20orf30; dJ1116H23.2.1; HSPC274; HSPC274 protein; RGD1307399; TMEM230; Transmembrane protein 230; UNQ2432/PRO4992; UPF0414 transmembrane protein C20orf30; UPF0414 transmembrane protein C20orf30 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human LRRC25 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126364
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.