Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983465
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3465 Supplier Bioss Supplier No. BS15111R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AAR2 encodes the homolog of the yeast A1-alpha2 repressin protein that is involved in mRNA splicing. Alternately spliced transcript variants have been found for this gene.

Specifications

Antigen C20orf4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AAR2
Gene Accession No. Q9Y312
Gene Alias 0610011L14Rik; Aar2; AAR2 splicing factor homolog; AAR2 splicing factor homolog (S. cerevisiae); C20orf4; CGI-23; novel transcript; PRO0225; Protein AAR2 homolog; RGD1311066
Gene Symbols AAR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25980
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.