Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf43 Monoclonal Antibody (OTI7E10), TrueMAB™, OriGene
SDP

Catalog No. 502541558
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1558 30 μL
50-167-3482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1558 Supplier OriGene Supplier No. TA502421S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C20orf43 Monoclonal antibody specifically detects C20orf43 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen C20orf43
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI7E10
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MSI1
Gene Accession No. O43347
Gene Alias 1700067C04Rik; 2410001C21Rik; 5730427M17Rik; AA589417; AD-007; C20orf43; CDA05; CDAO5; HSPC164; HSPC169; likely orthologue of H. sapiens chromosome 20 open reading frame 43 (C20orf43); protein RTF2 homolog; Replication termination factor 2; replication termination factor 2 domain containing 1; replication termination factor 2 domain-containing protein 1; RGD1311072; Rtf2; RTFDC1; SHUJUN-3; UPF0549 protein C20orf43; UPF0549 protein C20orf43 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human MSI1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4440
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.