Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C20orf43 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546767
Description
Peptide sequence: EKALGKAASH IKSIKNVTEL KLSDNPAWEG DKGNTKGDKH DDLQRARFIC.
C20orf43 protein function remains unknown.
Specifications
C20orf43 | |
Polyclonal | |
Unconjugated | |
RTF2 | |
1700067C04Rik; 2410001C21Rik; 5730427M17Rik; AA589417; AD-007; C20orf43; CDA05; CDAO5; HSPC164; HSPC169; likely orthologue of H. sapiens chromosome 20 open reading frame 43 (C20orf43); protein RTF2 homolog; Replication termination factor 2; replication termination factor 2 domain containing 1; replication termination factor 2 domain-containing protein 1; RGD1311072; Rtf2; RTFDC1; SHUJUN-3; UPF0549 protein C20orf43; UPF0549 protein C20orf43 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51507 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BY42 | |
RTF2 | |
Synthetic peptide directed towards the N-terminal of human RTFDC1 (aa 72-121). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction