Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C20orf43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562891 Supplier Invitrogen™ Supplier No. PA562891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AWEGDKGNTK GDKHDDLQRA RFICPVVGLE MNGRHRFCFL RCCGCVFSER ALKEIKAEVC HTCGAAFQED DVIVLNGTKE DVDVLK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

C20orf43 protein function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf43
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTF2
Gene Accession No. Q9BY42
Gene Alias 1700067C04Rik; 2410001C21Rik; 5730427M17Rik; AA589417; AD-007; C20orf43; CDA05; CDAO5; HSPC164; HSPC169; likely orthologue of H. sapiens chromosome 20 open reading frame 43 (C20orf43); protein RTF2 homolog; Replication termination factor 2; replication termination factor 2 domain containing 1; replication termination factor 2 domain-containing protein 1; RGD1311072; Rtf2; RTFDC1; SHUJUN-3; UPF0549 protein C20orf43; UPF0549 protein C20orf43 homolog
Gene Symbols RTF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RTFDC1. Recombinant protein control fragment (Product #RP-99289).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.