Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C20orf7 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15113R

Description
The NADH-ubiquinone oxidoreductase complex (complex I) of the mitochondrial respiratory chain catalyzes the transfer of electrons from NADH to ubiquinone, and consists of at least 43 subunits. The complex is located in the inner mitochondrial membrane. This gene encodes a mitochondrial protein that is associated with the matrix face of the mitochondrial inner membrane and is required for complex I assembly. A mutation in this gene results in mitochondrial complex I deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
C20orf7 | |
Polyclonal | |
Unconjugated | |
NDUFAF5 | |
2310003L22Rik; Arginine-hydroxylase NDUFAF5, mitochondrial; bA526K24.2; BOS_12779; C13H20orf7; C20orf7; C24H20orf7; CG8067-like; chromosome 13 open reading frame, human C20orf7; dJ842G6.1; LOC100337968; NADH dehydrogenase (ubiquinone) complex I, assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; NADH:ubiquinone oxidoreductase complex assembly factor 5; ndufaf5; probable methyltransferase C20orf7 homolog, mitochondrial; probable methyltransferase C20orf7, mitochondrial; Putative methyltransferase NDUFAF5; RGD1309829; zgc:162919 | |
Rabbit | |
Protein A | |
RUO | |
79133 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5TEU4 | |
NDUFAF5 | |
KLH conjugated synthetic peptide derived from human C20orf7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction