Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf70 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983467
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3467 Supplier Bioss Supplier No. BS15114R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPLUNC2 is one member of PLUNC family, the gene undergoes alternative splicing using two 5′ non-coding exons, suggesting that the gene is regulated by alternative promoters.multiple SPLUNC2 isoforms are found in the oral cavity and suggest that these proteins may be differentially regulated in distinct tissues where they may function in the innate immune response. Mucin plugs, mucous and intermediate cells of mucoepidermoid carcinomas were positive for LPLUNC1 and SPLUNC2, but areas composed of epidermoid and clear cells were negative for all PLUNCs. Papillary cystadenocarcinoma was positive for all PLUNCs.

Specifications

Antigen C20orf70
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BPIFA2
Gene Accession No. Q96DR5
Gene Alias bA49G10.1; BPI fold containing family A member 2; BPI fold containing family A, member 2; BPI fold containing family A, member 2E; BPI fold-containing family A member 2; Bpifa2; Bpifa2e; C20orf70; mSplunc2; Parotid secretory protein; Psp; Short palate, lung and nasal epithelium carcinoma-associated protein 2; SPLUNC2; UNQ510/PRO1025
Gene Symbols BPIFA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf70.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140683
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.