Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C20orf72 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113423
Description
C20orf72 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The function of this protein remains unknown.
Specifications
| C20orf72 | |
| Polyclonal | |
| Unconjugated | |
| MGME1 | |
| 8430406I07Rik; AI426476; bA504H3.4; C13H20orf72; C20orf72; DDK1; LOW QUALITY PROTEIN: mitochondrial genome maintenance exonuclease 1; MGME1; mitochondrial genome maintainance exonuclease 1; mitochondrial genome maintenance exonuclease 1; MTDPS11; RGD1307465 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 92667 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BQP7 | |
| MGME1 | |
| Recombinant Human Mitochondrial genome maintenance exonuclease 1 protein (81-260AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction