Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C20orf72 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118060
This item is not returnable.
View return policy
Description
C20orf72 Polyclonal Antibody for Western Blot, IP
The function of this protein remains unknown.
Specifications
C20orf72 | |
Polyclonal | |
Unconjugated | |
MGME1 | |
8430406I07Rik; AI426476; bA504H3.4; C13H20orf72; C20orf72; DDK1; LOW QUALITY PROTEIN: mitochondrial genome maintenance exonuclease 1; MGME1; mitochondrial genome maintainance exonuclease 1; mitochondrial genome maintenance exonuclease 1; MTDPS11; RGD1307465 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
92667 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9BQP7 | |
MGME1 | |
E. coli-derived Human MGME1/C20orf72 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction