Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf79 Monoclonal Antibody (OTI2B5), TrueMAB™, OriGene
SDP

Catalog No. 501684698
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4698 Supplier OriGene Supplier No. TA812878
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SCP2D1 (SCP2 Sterol Binding Domain Containing 1) is a Protein Coding gene. Diseases associated with SCP2D1 include Corneal Dystrophy, Posterior Polymorphous, 1 and D-Bifunctional Protein Deficiency. An important paralog of this gene is HSD17B4.

Specifications

Antigen C20orf79
Applications Western Blot
Classification Monoclonal
Clone OTI2B5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SCP2D1
Gene Accession No. Q9UJQ7
Gene Alias C20orf79, HSD22, dJ1068E13.2
Gene Symbols SCP2D1
Host Species Mouse
Immunogen Full length human recombit protein of human C20orf79 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140856
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.