Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf79 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983471
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3471 Supplier Bioss Supplier No. BS15118R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCP2D1 (SCP2 Sterol Binding Domain Containing 1) is a Protein Coding gene. Diseases associated with SCP2D1 include Corneal Dystrophy, Posterior Polymorphous, 1 and D-Bifunctional Protein Deficiency. An important paralog of this gene is HSD17B4.

Specifications

Antigen C20orf79
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SCP2D1
Gene Accession No. Q9UJQ7
Gene Alias C20orf79; dJ1068E13.2; HSD22; SCP2 sterol binding domain containing 1; SCP2 sterol-binding domain containing 1; SCP2 sterol-binding domain-containing protein 1; SCP2D1; sterol carrier protein 2-like protein
Gene Symbols SCP2D1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf79.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140856
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.