Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C20orf94 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983472
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3472 Supplier Bioss Supplier No. BS15119R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLX4IP/C20orf94 interacts with SLX4/BTBD12.

Specifications

Antigen C20orf94
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Slx4ip
Gene Accession No. Q5VYV7
Gene Alias 2210009G21Rik; 2410004I22Rik; bA204H22.1; bA254M13.1; C20orf94; C85661; dJ1099D15.3; Protein SLX4IP; SLX4 interacting protein; SLX4-interacting protein; SLX4IP; Slxip; UPF0492 protein C20orf94 homolog
Gene Symbols Slx4ip
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C20orf94.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128710
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.