Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ C20S Compact Coulometric KF Titrators
GSA_VA

Catalog No. 01914313
Click to view available options
Includes:
Diaphragm
No Diaphragm

Ideal for fast and precise water content determination with very simple operation. Mettler Toledo C20S Coulometric KF Titrators can analyze samples with water content between 1ppm and 5% in single measurements or as sample series with statistics.

Karl Fischer Titration is the specific standard method for water content determination and gives accurate and precise results within minutes.

One Click™ User Interface

  • Large color touchscreen (VGA 5.7 in. color TFT, 640 x 480 pixel) with simple-to-understand and fast-to-operate One Click user interface
  • All users have their own homescreen with dedicated shortcut buttons (12 per user) to directly start specific routine tasks
  • User interface in English, German, French, Spanish, Italian, Chinese, Russian, Polish, Korean and Portuguese available

Diaphragmless Cell

  • Choice of a current generator cell with or without a diaphragm
  • Diaphragmless cell offers more convenience for most samples as there is no catholyte needed
  • Due to the unique design, the diaphragmless cell even meets the demands of very precise low water content determinations

Solvent Manager for Safe Handling

  • Protects users against contact with chemicals
  • Takes care of filling, draining and exchange of reagents
  • It is controlled by the titrator and full security is guaranteed by overflow protector in the waste bottle

PC software LabX™ Titration

  • Connectivity to LabX Titration PC software via USB or direct Ethernet (TCP/IP)
  • Full control over the titrator, secure database archiving, networking, efficient result management with control charts and individual search filters or rapid and clear method development for new applications
  • Complete traceability by providing user management, audit trail, method history and electronic signature

Flexible Data Export

  • Devices such as printers, memory sticks, USB hub or USB barcode readers can be connected via the USB port and are automatically recognized
  • Common USB printers (DIN-A4/US letter) or the USB-P25 compact printer for secure and GLP-compliant paper recording

Communication

  • Data Export: USB, Ethernet, PDF
  • Ethernet (TCP/IP): 1
  • COM (serial balance connection): 1
  • USB host (printer, memory stick, barcode reader, hub): 2
  • USB client (PC): 1
  • Integrated Magnetic Stirrer: 1
  • Pump/Stirrer: 1

Methods and Series

  • KF standard methods: 1
  • Max. number of user methods: 5
  • Number of method functions: 12
  • Number of samples per series: 120
  • Autostart: automatic start after sample addition: Yes
  • Max. number of calculations/results per method: 3
  • Statistics of up to 120 samples including result limits check, outlier test: Yes
  • Recalculation of results: Yes
  • Data in Online Screen: time, water count, potential, stirrer speed, consumption, drift: Yes
  • Data in measured value table: time, water count, potential, consumption, drift: Yes
  • Curves in Online Screen and printout: E-C, E-t, C-t, drift-t, water count-t: Yes

Application

  • Liquid and gaseous samples
  • Resources and method database

Specifications

  • Measurement Range: -2000 to +2000mV
  • Resolution/Limits of Error: 0.1mV/2mV
  • Polarization Current Range: 0 to 24μA
  • Polarization Current Resolution/Limits of Error: 0.1μA/1μA
  • Ambient Temperature: 5° to 40°C
  • Atmospheric Humidity: max. 80% at 31°C
  • Drift Measurement: Online, <2μg/min.
  • Measurement Range: 10μg to 200mg
  • Resolution: 0.1μg
  • Repeatability: 0.3% at >1mg water
  • Glass Beaker: 200mL
  • Dimensions (D x W x H): 34 x 21 x 31.2cm
  • Weight: 3.3kg

Certifications

CE, ISO 9001, ISO 14001

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, ISO 9001, ISO 14001
Detection Range 0 to 24μA (Polarization Current Range)
Display Type VGA 5.7 in. color TFT, 640 x 480 pixel
Type Coulometric KF Titrator
For Use With (Application) Liquid and gaseous samples; Resources and method database
Includes No Diaphragm
Model C20SX
Range (mV) -2000 to +2000 mV
Resolution 0.1mV/2mV; 0.1μA/1μA; 0.1μg
Sample Size 120 (per series)
Temperature Range (Metric) Ambient 5°C to 40°C
Width (Metric) 21 cm
Depth (Metric) 34 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.