Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C21orf140 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560932 Supplier Invitrogen™ Supplier No. PA560932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPLLRNVII RSQFDGIKRK QCLQYLKTLR TLQYDGFKTV YFGETNIPES LVTGEDISDG YFIQTPTWCI VHAAGSQGWV PWKYRVF Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf140
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C21orf140
Gene Accession No. B9A014
Gene Alias C21orf140; chromosome 21 open reading frame 140; FAM243A; FAM243B; Protein FAM243A; Protein FAM243B; uncharacterized protein C21orf140; uncharacterized protein ENSP00000386791
Gene Symbols C21orf140
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C21orf140. Recombinant protein control fragment (Product #RP-100357).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101928147
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.