Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C21orf2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140310 Supplier Invitrogen™ Supplier No. PA5140310
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PVSRCQRLSE LYLRRNRIPS LAELFYLKGL PRLRVLWLAE NPCCGTSPHR YRMTVLRTLP RLQKLDNQAV TEEELSRALS EGEEITAA Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 85%.

Four alternatively spliced transcript variants encoding four different isoforms have been found for this nuclear gene. All isoforms contain leucine-rich repeats. Three of these isoforms are mitochondrial proteins and one of them lacks the target peptide, so is not located in mitochondrion. This gene is down-regulated in Down syndrome (DS) brain, which may represent mitochondrial dysfunction in DS patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C21orf2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP410
Gene Accession No. O43822
Gene Alias C21orf2; C21orf-HUMF09G8.5; cDNA A2-YF5; CFAP410; chromosome 21 open reading frame 2; Cilia- and flagella-associated protein 410; cu002; leucine rich repeat containing 76; leucine-rich repeat-containing protein 76; LRRC76; nuclear encoded mitochondrial protein C21orf2; protein C21orf2; YF5; YF5/A2
Gene Symbols CFAP410
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CFAP410. Recombinant protein control fragment (Product #RP-108791).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.