Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf25 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989503
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9503 Supplier Bioss Supplier No. BS9988R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C21orf25
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C2CD2
Gene Accession No. Q9Y426
Gene Alias C2 calcium dependent domain containing 2; C2 calcium-dependent domain containing 2; C2 domain-containing protein 2; C21orf25; C21orf258; C2CD2; TMEM24 related geme; TMEM24L; Transmembrane protein 24-like
Gene Symbols C2CD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C2CD2/C21orf25.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25966
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.