Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf45 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735090 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5090 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5090 Supplier Proteintech Group Inc Supplier No. 258321AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C21orf45, also named as MIS18A, C21orf46 and FASP1, is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis. C21orf45 has dimer (∽52 kDa) and heterodimer(∽105 kDa) forms.

Specifications

Antigen C21orf45
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MIS18A
Gene Accession No. Q9CZJ6, Q9NYP9
Gene Alias B28, C21orf45, C21orf46, FAPP1 associated protein 1, FASP1, MIS18A, MIS18alpha, Protein Mis18 alpha
Gene Symbols MIS18A
Host Species Rabbit
Immunogen C21orf45 Fusion Protein Ag22922
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54069, 66578
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.