Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C21orf56 Monoclonal Antibody (OTI1H6), TrueMAB™, OriGene
SDP

Catalog No. 502540204
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0204 30 μL
50-167-5280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0204 Supplier OriGene Supplier No. TA504634S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C21orf56 Monoclonal antibody specifically detects C21orf56 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen C21orf56
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H6
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Sat2
Gene Accession No. Q96F10
Gene Alias 1700022B01Rik; 1700027D21Rik; AA062323; C1H21orf56; C21orf56; C31H21orf56; hypothetical protein LOC522100; Spatc1l; Speriolin-like protein; spermatogenesis and centriole associated 1 like; spermatogenesis and centriole associated 1-like; spermatogenesis and centriole-associated protein 1-like protein; uncharacterized protein C21orf56 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human SAT2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112483
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.