Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C21orf59 Antibody (OTI2H5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272060AF350
Description
C21orf59 Monoclonal specifically detects C21orf59 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
C21orf59 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CFAP298 | |
Full length recombinant protein of human C21orf59 (NP_067077) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2H5 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
C21orf48, chromosome 21 open reading frame 48, chromosome 21 open reading frame 59, FLJ20467, FLJ37137, FLJ40247, hypothetical protein LOC56683, kur, kurly | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
56683.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction