Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C21orf59 Monoclonal Antibody (OTI2B2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501386S


Description
C21orf59 Monoclonal antibody specifically detects C21orf59 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
C21orf59 | |
Monoclonal | |
0.54 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P00387 | |
Mouse | |
Affinity chromatography | |
RUO | |
1727 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2B2 | |
Unconjugated | |
CYB5R3 | |
C21orf48; C21orf59; CFAP298; chromosome 21 open reading frame 59; CILD26; Cilia- and flagella-associated protein 298; FBB18; Kur; prostate cancer upregulated protein 1; Protein kurly homolog; UPF0769 protein C21orf59 | |
Full length human recombinant protein of human CYB5R3 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction